accessibility menu, dialog, popup

UserName

CAS RN 664993-53-7

H3COOHNOHNOO

CAS RN 664993-53-7

IUPAC Name: N-[4-({[4-(4-methoxyphenyl)oxan-4-yl]methyl}carbamoyl)phenyl]furan-2-carboxamide
Synonyms: N-[4-({[4-(4-methoxyphenyl)oxan-4-yl]methyl}carbamoyl)phenyl]furan-2-carboxamide
Molecular Weight (g/mol): 434.49
Molecular Formula: C25H26N2O5
InChi Key: ZJZWZIXSGNFWQQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1(CNC(=O)C2=CC=C(NC(=O)C3=CC=CO3)C=C2)CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results