accessibility menu, dialog, popup

UserName

FFFFFFOPOOFFFFFFOFFFFFF

CAS RN 66489-68-7

IUPAC Name: tris(1,1,1,3,3,3-hexafluoropropan-2-yl) phosphate
Synonyms: tris(1,1,1,3,3,3-hexafluoropropan-2-yl) phosphate
Molecular Weight (g/mol): 548.06
Molecular Formula: C9H3F18O4P
InChi Key: QLCATRCPAOPBOP-UHFFFAOYSA-N
SMILES: FC(F)(F)C(OP(=O)(OC(C(F)(F)F)C(F)(F)F)OC(C(F)(F)F)C(F)(F)F)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results