missing translation for 'onlineSavingsMsg'
Learn More

Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphate 98.0+%, TCI America™

Catalog No. T30415G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30415G 5 g
T30411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30415G Supplier TCI America Supplier No. T30415G
Only null left

Chemical Identifiers

CAS 66489-68-7
Molecular Formula C9H3F18O4P
Molecular Weight (g/mol) 548.064
InChI Key QLCATRCPAOPBOP-UHFFFAOYSA-N
Synonym Phosphoric Acid Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Ester
PubChem CID 13681431
IUPAC Name tris(1,1,1,3,3,3-hexafluoropropan-2-yl) phosphate
SMILES C(C(F)(F)F)(C(F)(F)F)OP(=O)(OC(C(F)(F)F)C(F)(F)F)OC(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 32°C
Boiling Point 72°C
Quantity 5 g
Formula Weight 548.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphate
TSCA No
Recommended Storage Refrigerator