accessibility menu, dialog, popup

UserName

CH3OOCH3H3C(Z)

CAS RN 6622-76-0

IUPAC Name: methyl (2Z)-2-methylbut-2-enoate
Synonyms: methyl tiglate
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: YYJWBYNQJLBIGS-PLNGDYQASA-N
SMILES: COC(=O)C(\C)=C/C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)