accessibility menu, dialog, popup

UserName

CAS RN 66107-34-4

CH3OSOOFFF

CAS RN 66107-34-4

IUPAC Name: 2-methylphenyl trifluoromethanesulfonate
Synonyms: 2-methylphenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 240.2
Molecular Formula: C8H7F3O3S
InChi Key: RBOGJRXPKONDGC-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1OS(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results