accessibility menu, dialog, popup

UserName

CH3H3CNHCH3H3CHOOClCl

CAS RN 660-27-5

IUPAC Name: 2,2-dichloroacetic acid; bis(propan-2-yl)amine
Synonyms: dichloroacetic acid; diisopropylamine
Molecular Weight (g/mol): 230.13
Molecular Formula: C8H17Cl2NO2
InChi Key: ILKBHIBYKSHTKQ-UHFFFAOYSA-N
SMILES: OC(=O)C(Cl)Cl.CC(C)NC(C)C
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)