accessibility menu, dialog, popup

UserName

CAS RN 65880-17-3

H3COOHN

CAS RN 65880-17-3

IUPAC Name: ethyl 4,5,6,7-tetrahydro-2H-isoindole-1-carboxylate
Synonyms: ethyl 4,5,6,7-tetrahydro-2H-isoindole-1-carboxylate
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: NIMDFIORBNYIGB-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C2CCCCC2=CN1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results