missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,5,6,7-Tetrahydroisoindole-1-carboxylate 95.0+%, TCI America™

Catalog No. E08571G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E08571G Supplier TCI America Supplier No. E08571G
Only null left

Chemical Identifiers

CAS 65880-17-3
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD06227773
InChI Key NIMDFIORBNYIGB-UHFFFAOYSA-N
Synonym 4,5,6,7-Tetrahydroisoindole-1-carboxylic Acid Ethyl Ester
PubChem CID 10035498
IUPAC Name ethyl 4,5,6,7-tetrahydro-2H-isoindole-1-carboxylate
SMILES CCOC(=O)C1=C2CCCCC2=CN1

Specifications

Melting Point 79°C
Quantity 1 g
Formula Weight 193.25
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4,5,6,7-Tetrahydroisoindole-1-carboxylate
TSCA No
Recommended Storage Refrigerator