accessibility menu, dialog, popup

UserName

OOClCl

CAS RN 6575-24-2

IUPAC Name: 2-(2,6-dichlorophenyl)acetate
Synonyms: 2-(2,6-dichlorophenyl)acetate
Molecular Weight (g/mol): 204.03
Molecular Formula: C8H5Cl2O2
InChi Key: SFAILOOQFZNOAU-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=C(Cl)C=CC=C1Cl
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)