CAS RN 6575-05-9
IUPAC Name:
2,4,6-trichlorobenzonitrile
Synonyms:
2,4,6-trichlorobenzonitrile
Molecular Weight (g/mol):
206.45
Molecular Formula:
C7H2Cl3N
InChi Key:
PGODHCIOIPODFE-UHFFFAOYSA-N
SMILES:
ClC1=CC(Cl)=C(C#N)C(Cl)=C1
1
–
2
of
2
results
1
2
2,4,6-Trichlorobenzonitrile 98.0+%, TCI America™
CAS: 6575-05-9 Molecular Formula: C7H2Cl3N Molecular Weight (g/mol): 206.45 MDL Number: MFCD00052711 InChI Key: PGODHCIOIPODFE-UHFFFAOYSA-N Synonym: benzonitrile, 2,4,6-trichloro,2,4,6-trichlorobenzenecarbonitrile,pubchem4822,acmc-1b2tu,pgodhcioipodfe-uhfffaoysa PubChem CID: 522179 IUPAC Name: 2,4,6-trichlorobenzonitrile SMILES: C1=C(C=C(C(=C1Cl)C#N)Cl)Cl