accessibility menu, dialog, popup

UserName

CAS RN 65610-14-2

FNN

CAS RN 65610-14-2

IUPAC Name: 4-fluorobenzene-1,2-dicarbonitrile
Synonyms: 4-fluorobenzene-1,2-dicarbonitrile
Molecular Weight (g/mol): 146.12
Molecular Formula: C8H3FN2
InChi Key: QQEKYCCJLSRLEC-UHFFFAOYSA-N
SMILES: FC1=CC=C(C#N)C(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results