accessibility menu, dialog, popup

UserName

H3COOHNNH

CAS RN 64951-36-6

IUPAC Name: ethyl N-(piperidin-4-yl)carbamate
Synonyms: ethyl N-(piperidin-4-yl)carbamate
Molecular Weight (g/mol): 172.23
Molecular Formula: C8H16N2O2
InChi Key: JIRHHWNUZXRDHN-UHFFFAOYSA-N
SMILES: CCOC(=O)NC1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)