missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethoxycarbonylamino)piperidine, 97%

Catalog No. AAH6309506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6309506 5 g
AAH6309503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6309506 Supplier Thermo Scientific Chemicals Supplier No. H6309506
Only null left

CAS: 64951-36-6 | C8H16N2O2 | 172.228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64951-36-6
Molecular Formula C8H16N2O2
Molecular Weight (g/mol) 172.228
MDL Number MFCD03844679
InChI Key JIRHHWNUZXRDHN-UHFFFAOYSA-N
Synonym ethyl piperidin-4-ylcarbamate, ethyl 4-piperidylcarbamate, 4-ethoxycarbonylaminopiperidine, 4-ethoxycarbonylamino piperidine, ethyl n-piperidin-4-yl carbamate, carbamic acid, n-4-piperidinyl-, ethyl ester
PubChem CID 15595030
IUPAC Name ethyl N-piperidin-4-ylcarbamate
SMILES CCOC(=O)NC1CCNCC1

Specifications

Quantity 5 g
Formula Weight 172.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(Ethoxycarbonylamino)piperidine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only