missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Cyclohexene-1-carboxylate 98.0+%, TCI America™

Catalog No. C0967500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0967500ML 500 mL
C096725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0967500ML Supplier TCI America Supplier No. C0967500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6493-77-2
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00053266
InChI Key IPUNVLFESXFVFH-UHFFFAOYNA-N
Synonym methyl cyclohex-3-enecarboxylate, 3-cyclohexene-1-carboxylic acid methyl ester, methyl 3-cyclohexenecarboxylate, 3-cyclohexene-1-carboxylic acid, methyl ester, methyl 3-cyclohexene-1-carboxylate, 1-carbomethoxy-3-cyclohexene, methyl 4-cyclohexenecarboxylate, methyl 1,2,3,6-tetrahydrobenzoate, 1,3-butadiene-methyl acrylate adduct, acmc-1bity
PubChem CID 96926
IUPAC Name methyl cyclohex-3-ene-1-carboxylate
SMILES COC(=O)C1CCC=CC1

Specifications

Color Yellow
Boiling Point 185°C
Quantity 500 mL
UN Number 3272
Formula Weight 140.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Cyclohexene-1-carboxylate