accessibility menu, dialog, popup

UserName

CH3NCH3SOOCH3

CAS RN 649-15-0

IUPAC Name: N,N-diethyl-4-methylbenzene-1-sulfonamide
Synonyms: N,N-diethyl-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 227.32
Molecular Formula: C11H17NO2S
InChi Key: AOJBACHWNDMRQP-UHFFFAOYSA-N
SMILES: CCN(CC)S(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results