missing translation for 'onlineSavingsMsg'
Learn More

Tetracosane 99.0+%, TCI America™

Catalog No. T0075500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0075500G 500 g
T007525G 25 g
T0075100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0075500G Supplier TCI America Supplier No. T0075500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 646-31-1
Molecular Formula C24H50
Molecular Weight (g/mol) 338.664
MDL Number MFCD00009352
InChI Key POOSGDOYLQNASK-UHFFFAOYSA-N
Synonym n-tetracosane, lignocerane, unii-yq5h1m1d7i, tricosane, methyl, yq5h1m1d7i, tetracosane, analytical standard, tetracosane, acmc-1b0gq, n-tetracosane 5g, ksc357o9b
PubChem CID 12592
ChEBI CHEBI:32936
IUPAC Name tetracosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 51°C
Color White
Boiling Point 250°C
Quantity 500 g
Formula Weight 338.66
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Tetracosane
EINECSNumber (2)-0010
TSCA Yes