accessibility menu, dialog, popup

UserName

N

CAS RN 645-59-0

IUPAC Name: 3-phenylpropanenitrile
Synonyms: benzylacetonitrile
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: ACRWYXSKEHUQDB-UHFFFAOYSA-N
SMILES: N#CCCC1=CC=CC=C1
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)