missing translation for 'onlineSavingsMsg'
Learn More

Hydrocinnamonitrile, 99+%

Catalog No. AC173990100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173990100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC173990100 Supplier Thermo Scientific Chemicals Supplier No. 173990100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 645-59-0 | C9H9N | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 645-59-0
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00001961
InChI Key ACRWYXSKEHUQDB-UHFFFAOYSA-N
Synonym 3-phenylpropionitrile, benzenepropanenitrile, hydrocinnamonitrile, 2-phenylethyl cyanide, benzenepropionitrile, 2-cyanoethyl benzene, 3-phenylpropiononitrile, phenylpropionitrile, phenethyl cyanide, hydrocinnamique nitrile
PubChem CID 12581
ChEBI CHEBI:85426
IUPAC Name 3-phenylpropanenitrile
SMILES C1=CC=C(C=C1)CCC#N

Specifications

Melting Point -2°C to 1°C
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 261°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.5210 to 1.523
Linear Formula C6H5CH2CH2CN
Quantity 10 g
Beilstein 09, 512
Specific Gravity 1
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol,ethyl ether,slightly in chloroform
Formula Weight 131.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Hydrocinnamonitrile, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-447-8

RUO – Research Use Only