missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropionitrile 98.0+%, TCI America™

Catalog No. P061125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P061125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P061125G Supplier TCI America Supplier No. P061125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 645-59-0
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00001961
InChI Key ACRWYXSKEHUQDB-UHFFFAOYSA-N
Synonym 3-phenylpropionitrile, benzenepropanenitrile, hydrocinnamonitrile, 2-phenylethyl cyanide, benzenepropionitrile, 2-cyanoethyl benzene, 3-phenylpropiononitrile, phenylpropionitrile, phenethyl cyanide, hydrocinnamique nitrile
PubChem CID 12581
ChEBI CHEBI:85426
IUPAC Name 3-phenylpropanenitrile
SMILES C1=CC=C(C=C1)CCC#N

Specifications

Color Yellow
Boiling Point 254°C
Quantity 25 g
UN Number 3276
Formula Weight 131.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Phenylpropionitrile
EINECSNumber (3)-3119
RTECSNumber MW5604750
TSCA Yes