accessibility menu, dialog, popup

UserName

H3CONH2OCH3

CAS RN 645-36-3

IUPAC Name: 2,2-diethoxyethan-1-amine
Synonyms: ethanamine, 2,2-diethoxy-
Molecular Weight (g/mol): 133.19
Molecular Formula: C6H15NO2
InChi Key: HJKLEAOXCZIMPI-UHFFFAOYSA-N
SMILES: CCOC(CN)OCC
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)