missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetaldehyde diethyl acetal, 98%

Catalog No. AAA1042722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1042722 100 g
AAA1042714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1042722 Supplier Thermo Scientific Chemicals Supplier No. A1042722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 645-36-3 | C6H15NO2 | 133.19 g/mol

Aminoacetaldehyde diethyl acetal, is used as an agrochemical intermediate, pharmaceutical, syntheses material intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminoacetaldehyde diethyl acetal, is used as an agrochemical intermediate, pharmaceutical, syntheses material intermediate.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Store in a cool, dry place. Store in a tightly closed container.Take measures to prevent the build up of electrostatic charge. Stable under recommended storage conditions. Incompatabile to strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, Carbon dioxide (CO2).

Chemical Identifiers

CAS 645-36-3
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00008136
InChI Key HJKLEAOXCZIMPI-UHFFFAOYSA-N
Synonym 2,2-diethoxyethylamine, aminoacetaldehyde diethyl acetal, ethanamine, 2,2-diethoxy, 2,2-diethoxyethan-1-amine, 2-aminoacetaldehyde diethyl acetal, glycinaldehyde diethyl acetal, aminoacetal, diethoxyethylamine, acetaldehyde, amino-, diethyl acetal, aminoacetaldehydediethylacetal
PubChem CID 69524
IUPAC Name 2,2-diethoxyethanamine
SMILES CCOC(CN)OCC

Specifications

Density 0.916
Boiling Point 162°C to 163°C
Flash Point 45°C (113°F)
Refractive Index 1.417
Linear Formula H2NCH2CH(OCH2CH3)2
Quantity 100 g
UN Number UN2733
Beilstein 506067
Solubility Information Soluble in water.
Formula Weight 133.19
Percent Purity 98%
Chemical Name or Material Aminoacetaldehyde diethyl acetal
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H228-H314-H318
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 211-439-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only