missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetal 97.0+%, TCI America™

Catalog No. A024725ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A024725ML 25 mL
A0247250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A024725ML Supplier TCI America Supplier No. A024725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 645-36-3
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00008136
InChI Key HJKLEAOXCZIMPI-UHFFFAOYSA-N
Synonym 2,2-diethoxyethylamine, aminoacetaldehyde diethyl acetal, ethanamine, 2,2-diethoxy, 2,2-diethoxyethan-1-amine, 2-aminoacetaldehyde diethyl acetal, glycinaldehyde diethyl acetal, aminoacetal, diethoxyethylamine, acetaldehyde, amino-, diethyl acetal, aminoacetaldehydediethylacetal
PubChem CID 69524
IUPAC Name 2,2-diethoxyethan-1-amine
SMILES CCOC(CN)OCC

Specifications

Color Yellow
Boiling Point 163°C
Quantity 25 mL
UN Number 2924
Formula Weight 133.19
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Aminoacetal