accessibility menu, dialog, popup

UserName

NOONOOFFFFFF

CAS RN 64465-34-5

IUPAC Name: 1-[1,1,1,3,3,3-hexafluoro-2-(3-nitrophenyl)propan-2-yl]-3-nitrobenzene
Synonyms: 1-[1,1,1,3,3,3-hexafluoro-2-(3-nitrophenyl)propan-2-yl]-3-nitrobenzene
Molecular Weight (g/mol): 394.23
Molecular Formula: C15H8F6N2O4
InChi Key: YETXOTPYXAYEMI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)C(C1=CC(=CC=C1)[N+]([O-])=O)(C(F)(F)F)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results