accessibility menu, dialog, popup

UserName

FFN

CAS RN 64248-64-2

IUPAC Name: 2,5-difluorobenzonitrile
Synonyms: 2,5-difluorobenzonitrile
Molecular Weight (g/mol): 139.11
Molecular Formula: C7H3F2N
InChi Key: OJTMHIMQUQOLJV-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(=C1)C#N
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)