missing translation for 'onlineSavingsMsg'
Learn More

Nuclear Fast Red Solution, Ricca Chemical

Catalog No. R5463200500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R5463200500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R5463200500 Supplier Ricca Chemical Company Supplier No. R5463200500A

Specifications

Chemical Name or Material Nuclear Fast Red Solution
CAS 6409-77-4, 7732-18-5, 7784-31-8
CAS Min % 93.0, 4.67, 0.09
CAS Max % 97.0, 4.86, 0.1
Grade Indicator
Odor Odorless
Packaging Natural Poly Bottle
Color Purple
pH 3
Quantity 500 mL
Physical Form Liquid
Show More Show Less