missing translation for 'onlineSavingsMsg'
Learn More

Nuclear Fast Red, TCI America™

Catalog No. N01841G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N01841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N01841G Supplier TCI America Supplier No. N01841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6409-77-4
Molecular Formula C14H11NNaO7S
Molecular Weight (g/mol) 360.292
MDL Number MFCD00078493
InChI Key KDEATZKCHKVXGA-UHFFFAOYSA-N
Synonym Kernechtrot, NFR
PubChem CID 131675027
IUPAC Name 4-amino-1,3-dihydroxy-9,10-dioxoanthracene-2-sulfonic acid;molecular hydrogen;sodium
SMILES [HH].C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C(=C3N)O)S(=O)(=O)O)O.[Na]

Specifications

Quantity 1 g
Physical Form Crystal/Powder
Formula Weight 357.27
Chemical Name or Material Nuclear Fast Red