accessibility menu, dialog, popup

UserName

H2OH3CONNH2NH2H3COHOHO

CAS RN 6402-23-9

IUPAC Name: 2-hydroxypropanoic acid 7-ethoxyacridine-3,9-diamine hydrate
Synonyms: 6,9-diamino-2-ethoxyacridine lactic acid hydrate
Molecular Weight (g/mol): 361.4
Molecular Formula: C18H23N3O5
InChi Key: NYEPHMYJRNWPLA-UHFFFAOYNA-N
SMILES: O.CC(O)C(O)=O.CCOC1=CC=C2N=C3C=C(N)C=CC3=C(N)C2=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)