missing translation for 'onlineSavingsMsg'
Learn More

6,9-Diamino-2-ethoxyacridine lactate monohydrate, 95%, Thermo Scientific Chemicals

Catalog No. AC406381000
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC406381000 100 g Glass bottle
AC406380050 5 g Glass bottle
AC406380100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC406381000 Supplier Thermo Scientific Chemicals Supplier No. 406381000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6402-23-9 | C3H6O3·H2O | 361.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6402-23-9 , 1837-57-6
Molecular Formula C3H6O3·H2O
Molecular Weight (g/mol) 361.39
MDL Number MFCD00149646
InChI Key NYEPHMYJRNWPLA-UHFFFAOYSA-N
Synonym ethacridine lactate monohydrate, ethodin, ethacridine lactate hydrate, ethacridine lactate salt, dsstox_cid_26274, dsstox_rid_81501, dsstox_gsid_46274, 7-ethoxyacridine-3,9-diamine; 2-hydroxypropanoic acid; hydrate, rivanol monohydrate
PubChem CID 165457
IUPAC Name 7-ethoxyacridine-3,9-diamine;2-hydroxypropanoic acid;hydrate
SMILES CCOC1=CC2=C(C3=C(C=C(C=C3)N)N=C2C=C1)N.CC(C(=O)O)O.O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 243°C to 245°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 3771
Solubility Information Solubility in water: .. Other solubilities: slowly soluble in 15 parts water,soluble in 9 parts boiling water,soluble in 110 parts alcohol (22°C),soluble in 100 parts alcohol (20°C)
Formula Weight 361.39
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 6, 9-Diamino-2-ethoxyacridine lactate monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only