missing translation for 'onlineSavingsMsg'
Learn More

Acrinol Monohydrate 98.0+%, TCI America™

Catalog No. A013625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A013625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A013625G Supplier TCI America Supplier No. A013625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6402-23-9
Molecular Formula C18H23N3O5
Molecular Weight (g/mol) 361.398
MDL Number MFCD00149646
InChI Key NYEPHMYJRNWPLA-UHFFFAOYSA-N
Synonym ethacridine lactate monohydrate, ethodin, ethacridine lactate hydrate, ethacridine lactate salt, dsstox_cid_26274, dsstox_rid_81501, dsstox_gsid_46274, 7-ethoxyacridine-3,9-diamine; 2-hydroxypropanoic acid; hydrate, rivanol monohydrate
PubChem CID 165457
IUPAC Name 7-ethoxyacridine-3,9-diamine;2-hydroxypropanoic acid;hydrate
SMILES CCOC1=CC2=C(C3=C(C=C(C=C3)N)N=C2C=C1)N.CC(C(=O)O)O.O

Specifications

Melting Point 245°C
Color Yellow
Quantity 25 g
Formula Weight 343.38
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Acrinol Monohydrate
RTECSNumber OD4725000
TSCA Yes