CAS RN 640-60-8
IUPAC Name:
phenyl 4-methylbenzene-1-sulfonate
Synonyms:
phenyl 4-methylbenzenesulfonate
Molecular Weight (g/mol):
248.3
Molecular Formula:
C13H12O3S
InChi Key:
KZQFPRKQBWRRHQ-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)S(=O)(=O)OC1=CC=CC=C1
1
–
1
of
1
results
1
Phenyl p-Toluenesulfonate 99.0+%, TCI America™
CAS: 640-60-8 Molecular Formula: C13H12O3S Molecular Weight (g/mol): 248.296 MDL Number: MFCD00025996 InChI Key: KZQFPRKQBWRRHQ-UHFFFAOYSA-N Synonym: phenyl para-toluenesulfonate,p-toluenesulfonic acid phenyl ester,phenyl tosylate,phenyl p-toluenesulfonate,phenylp-toluenesulfonate,acmc-209njz,phenyl 4-methylbenzene-1-sulfonate,toluene-4-sulfonic acid phenyl ester,benzenesulfonic acid,4-methyl-, phenyl ester PubChem CID: 223146 IUPAC Name: phenyl 4-methylbenzenesulfonate SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC2=CC=CC=C2