accessibility menu, dialog, popup

UserName

CH3H2N(R)OOOO

CAS RN 6384-92-5

IUPAC Name: (2R)-2-(methylazaniumyl)butanedioate
Synonyms: N-methyl-D-aspartic acid
Molecular Weight (g/mol): 146.12
Molecular Formula: C5H8NO4
InChi Key: HOKKHZGPKSLGJE-GSVOUGTGSA-M
SMILES: C[NH2+][C@H](CC([O-])=O)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)