accessibility menu, dialog, popup

UserName

H3COOOOHNH2

CAS RN 6384-08-3

IUPAC Name: 4-amino-5-methoxy-5-oxopentanoic acid
Synonyms: 4-amino-5-methoxy-5-oxopentanoic acid
Molecular Weight (g/mol): 161.16
Molecular Formula: C6H11NO4
InChi Key: SEWIYICDCVPBEW-UHFFFAOYNA-N
SMILES: COC(=O)C(N)CCC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)