accessibility menu, dialog, popup

UserName

H3CClO

CAS RN 638-29-9

IUPAC Name: pentanoyl chloride
Synonyms: pentanoyl chloride
Molecular Weight (g/mol): 120.58
Molecular Formula: C5H9ClO
InChi Key: XGISHOFUAFNYQF-UHFFFAOYSA-N
SMILES: CCCCC(Cl)=O
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)