missing translation for 'onlineSavingsMsg'
Learn More

Valeryl chloride, 98%

Catalog No. AC169121000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169121000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC169121000 Supplier Thermo Scientific Chemicals Supplier No. 169121000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 638-29-9 | C5H9ClO | 120.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 638-29-9
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD00000757
InChI Key XGISHOFUAFNYQF-UHFFFAOYSA-N
Synonym valeryl chloride, valeroyl chloride, valerylchloride, n-valeryl chloride, n-pentanoyl chloride, n-valeroyl chloride, pentanoic acid chloride, n-pentoyl chloride, valerylchlorid, valeroylchloride
PubChem CID 61186
IUPAC Name pentanoyl chloride
SMILES CCCCC(Cl)=O

Specifications

Melting Point -110.0°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 125.0°C to 127.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4190 to 1.4210
Linear Formula CH3(CH2)3COCl
Quantity 100 g
Beilstein 02,301
Specific Gravity 0.99
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in most common organic solvents
Formula Weight 120.58
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Valeryl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously
EINECSNumber 211-330-1
TSCA TSCA

RUO – Research Use Only