missing translation for 'onlineSavingsMsg'
Learn More

Valeryl chloride, 98%, Thermo Scientific™

Catalog No. AC169120010
Change view
Click to view available options
1kg, Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC169120010 1kg, Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC169120010 Supplier Thermo Scientific Chemicals Supplier No. 169120010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 638-29-9
Density 0.9900g/mL
Boiling Point 125.0°C to 127.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H9ClO
Refractive Index 1.4190 to 1.4210
Linear Formula CH3(CH2)3COCl
Quantity 1kg
MDL Number MFCD00000757
Beilstein 02,301
Specific Gravity 0.99
Synonym valeryl chloride, valeroyl chloride, valerylchloride, n-valeryl chloride, n-pentanoyl chloride, n-valeroyl chloride, pentanoic acid chloride, n-pentoyl chloride, valerylchlorid, valeroylchloride
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in most common organic solvents
InChI Key XGISHOFUAFNYQF-UHFFFAOYSA-N
SMILES CCCCC(Cl)=O
IUPAC Name pentanoyl chloride
Molecular Weight (g/mol) 120.58
PubChem CID 61186
Formula Weight 120.58
Percent Purity 98%
Chemical Name or Material Valeryl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously
EINECSNumber 211-330-1
TSCA TSCA

RUO – Research Use Only