accessibility menu, dialog, popup

UserName

H3CCH3Cl

CAS RN 638-28-8

IUPAC Name: 2-chlorohexane
Synonyms: 2-chlorohexane
Molecular Weight (g/mol): 120.62
Molecular Formula: C6H13Cl
InChi Key: GLCIPJOIEVLTPR-UHFFFAOYNA-N
SMILES: CCCCC(C)Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results