missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorohexane 98.0+%, TCI America™

Catalog No. C08905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08905G 5 g
C089025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C08905G Supplier TCI America Supplier No. C08905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 638-28-8
Molecular Formula C6H13Cl
Molecular Weight (g/mol) 120.62
MDL Number MFCD00039351
InChI Key GLCIPJOIEVLTPR-UHFFFAOYNA-N
Synonym 2-Hexyl Chloride
PubChem CID 12521
IUPAC Name 2-chlorohexane
SMILES CCCCC(C)Cl

Specifications

Color Colorless
Boiling Point 122°C
Quantity 5 g
UN Number 1993
Formula Weight 120.62
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorohexane
TSCA Yes
Recommended Storage Refrigerator