accessibility menu, dialog, popup

UserName

NH2(R)SHOOOHO

CAS RN 638-23-3

IUPAC Name: (2R)-2-amino-3-[(carboxymethyl)sulfanyl]propanoic acid
Synonyms: carboxymethylated cysteine mucojet sirup siroxyl superthiol sirup mucodyne carbocisteinum thiodril bronchokod mucocis mucolex sirup
Molecular Weight (g/mol): 179.19
Molecular Formula: C5H9NO4S
InChi Key: GBFLZEXEOZUWRN-VKHMYHEASA-N
SMILES: N[C@@H](CSCC(O)=O)C(O)=O
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)