missing translation for 'onlineSavingsMsg'
Learn More

S-(Carboxymethyl)-L-cysteine 98.0+%, TCI America™

Catalog No. C086325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C086325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C086325G Supplier TCI America Supplier No. C086325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity >98.0% S-(Carboxymethyl)-L-cysteine reagent for laboratory research applications.

Chemical Identifiers

CAS 638-23-3
Molecular Formula C5H9NO4S
Molecular Weight (g/mol) 179.19
MDL Number MFCD00002614
InChI Key GBFLZEXEOZUWRN-VKHMYHEASA-N
Synonym s-carboxymethyl-l-cysteine, carbocisteine, carbocysteine, carbocistein, transbronchin, bronchokod, lisomucil, mucolase, mucotab
PubChem CID 193653
ChEBI CHEBI:16163
IUPAC Name (2R)-2-amino-3-[(carboxymethyl)sulfanyl]propanoic acid
SMILES N[C@@H](CSCC(O)=O)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 179.19
Percent Purity ≥98.0% (HPLC,T)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material S-(Carboxymethyl)-L-cysteine
EINECSNumber (9)-1593
RTECSNumber AY4342000
TSCA Yes