missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-Carboxymethyl-L-cysteine, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 205.0°C to 207.0°C |
| Color | White |
| Ignition Residue | 0.2% max. |
| Loss on Drying | 0.5% max. (105°C, 3 hrs) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass bottle |
| Linear Formula | HO2CCH2SCH2CH(NH2)CO2H |
| Quantity | 100 g |
| Merck Index | 15, 1802 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap a |
| EINECSNumber | 211-327-5 |
| RTECSNumber | AY4342000 |
| TSCA | TSCA |
RUO – Research Use Only