accessibility menu, dialog, popup

UserName

CAS RN 637-81-0

H3COONNN

CAS RN 637-81-0

IUPAC Name: ethyl 2-azidoacetate
Synonyms: ethyl 2-azidoacetate
Molecular Weight (g/mol): 129.12
Molecular Formula: C4H7N3O2
InChi Key: HVJJYOAPXBPQQV-UHFFFAOYSA-N
SMILES: CCOC(=O)CN=[N+]=[N-]
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results