missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Azidoacetate 95.0+%, TCI America™

Catalog No. p-7129414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12555G 5 g
E125525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12555G Supplier TCI America Supplier No. E12555G
Only null left

Chemical Identifiers

CAS 637-81-0
Molecular Formula C4H7N3O2
Molecular Weight (g/mol) 129.119
MDL Number MFCD00190177
InChI Key HVJJYOAPXBPQQV-UHFFFAOYSA-N
Synonym ethyl azidoacetate, azidoacetic acid ethyl ester, acetic acid, azido-, ethyl ester, azido-acetic acid ethyl ester, ccris 3368, acetic acid, 2-azido-,ethyl ester, ethyl azido-acetate, ethyl2-azidoacetate, ethyl-2-azidoacetate, ethylazidoacetatesolution
PubChem CID 69480
IUPAC Name ethyl 2-azidoacetate
SMILES CCOC(=O)CN=[N+]=[N-]

Specifications

Color Yellow
Boiling Point 164°C
Quantity 5 g
UN Number 1993
Formula Weight 129.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Azidoacetate
TSCA No
Recommended Storage Refrigerator