accessibility menu, dialog, popup

UserName

HOOOHOHO

CAS RN 636-94-2

IUPAC Name: 2-hydroxybenzene-1,4-dicarboxylic acid
Synonyms: 2-hydroxybenzene-1,4-dicarboxylic acid
Molecular Weight (g/mol): 182.13
Molecular Formula: C8H6O5
InChi Key: CDOWNLMZVKJRSC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C(O)=O)C(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results