accessibility menu, dialog, popup

UserName

OHNONHONH2

CAS RN 63428-83-1

IUPAC Name: 6-amino-N-{5-[(6-oxohexyl)carbamoyl]pentyl}hexanamide
Synonyms: 6-amino-N-{5-[(6-oxohexyl)carbamoyl]pentyl}hexanamide
Molecular Weight (g/mol): 341.5
Molecular Formula: C18H35N3O3
InChi Key: HKJOYARIOKGODS-UHFFFAOYSA-N
SMILES: NCCCCCC(=O)NCCCCCC(=O)NCCCCCC=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results