accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
ICN09602
Structure Search
Print
Polyamide, MP Biomedicals™
Catalog No.
ICN09602
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
ICN09602
250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No.
ICN09602
Supplier
MP Biomedicals Inc
Supplier No.
0209602
Sign In
or
Register
to check your price and availability.
Quantity
Only null left
Add to cart
Add to cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Chemical Identifiers
CAS
63428-83-1
Molecular Formula
C18H35N3O3
Molecular Weight (g/mol)
341.50
InChI Key
HKJOYARIOKGODS-UHFFFAOYSA-N
Synonym
polyamide, polyamide without fluorescence
PubChem CID
71311215
Specifications
Quantity
250 g
Product Suggestions
Documents