accessibility menu, dialog, popup

UserName

NH2NHO

CAS RN 6320-39-4

IUPAC Name: 3-aminopyridin-4-ol
Synonyms: 3-aminopyridin-4-ol
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: OIIBRAGQGFLUFI-UHFFFAOYSA-N
SMILES: NC1=CN=CC=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results