missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-hydroxypyridine 98.0+%, TCI America™

Catalog No. A24325G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24325G 5 g
A24321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24325G Supplier TCI America Supplier No. A24325G

Chemical Identifiers

CAS 6320-39-4
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD04114230
InChI Key OIIBRAGQGFLUFI-UHFFFAOYSA-N
Synonym 3-amino-4-hydroxypyridine, 3-aminopyridin-4-ol, 3-amino-pyridin-4-ol, 4-pyridinol, 3-amino, 3-amino-4-pyridinol, 3-aminopyridin-4 1h-one, 4-hydroxy-3-aminopyridine, 3-amino-4-hydroxypyridine 4-hydroxy-3-amino pyridine, zlchem 466, acmc-209ndv
PubChem CID 233392
IUPAC Name 3-aminopyridin-4-ol
SMILES NC1=CN=CC=C1O

Specifications

Melting Point 150°C
Quantity 5 g
Formula Weight 110.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-hydroxypyridine