accessibility menu, dialog, popup

UserName

CAS RN 632-79-1

IUPAC Name: tetrabromo-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: tetrabromo-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 463.7
Molecular Formula: C8Br4O3
InChi Key: QHWKHLYUUZGSCW-UHFFFAOYSA-N
SMILES: BrC1=C2C(=O)OC(=O)C2=C(Br)C(Br)=C1Br
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)