missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylsilane 97.0+%, TCI America™

Catalog No. T15525ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15525ML 5 mL
T155225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15525ML Supplier TCI America Supplier No. T15525ML
Only null left

Chemical Identifiers

CAS 631-36-7
Molecular Formula C8H20Si
Molecular Weight (g/mol) 144.333
MDL Number MFCD00009019
InChI Key VCZQFJFZMMALHB-UHFFFAOYSA-N
Synonym silane, tetraethyl, tetraethylsilicon, tetraethylsilane, acmc-209ndb, c2h5 4si
PubChem CID 12426
IUPAC Name tetraethylsilane
SMILES CC[Si](CC)(CC)CC

Specifications

Color Colorless
Boiling Point 155°C
Quantity 5 mL
UN Number 1993
Formula Weight 144.33
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetraethylsilane
RTECSNumber VV5500000
TSCA Yes