accessibility menu, dialog, popup

UserName

H3CH3CCH3(S)OOCH3NH2HCl

CAS RN 63038-27-7

IUPAC Name: methyl (2S)-2-amino-3,3-dimethylbutanoate hydrochloride
Synonyms: methyl (2S)-2-amino-3,3-dimethylbutanoate hydrochloride
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: HRTQWUHFSXVRPY-XZNNNFJINA-N
SMILES: Cl.COC(=O)[C@@H](N)C(C)(C)C
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)